Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CMC Rescue™ Riggers Gloves
SDP

A synthetic leather palm combines the dexterity of a lighter glove, with the protection of a heavier leather glove. The pre-curved, anatomic design ensures a close and comfortable fit for working with knots or hardware.

$47.30

Specifications

Back Material Mesh
Color Black
For Use With (Application) Belay Systems, Power tool Operation, Main Line Hauling, and Repelling
Glove Cuff Style Elasticated
Material Synthetic Leather
View More Specs

Products 4
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
19-100-334
SDP
View Documents
CMC Rescue™
250302
Small
Pair for $47.30
Dozen of 12 PR for $567.50
Add to cart
 
19-100-335
SDP
View Documents
CMC Rescue™
250303
Medium
Pair for $47.30
Dozen of 12 PR for $567.50
Add to cart
 
19-100-336
SDP
View Documents
CMC Rescue™
250304
Large
Pair for $47.30
Dozen of 12 PR for $567.50
Add to cart
 
19-100-337
SDP
View Documents
CMC Rescue™
250305
X-Large
Dozen of 12 for $47.30
Add to cart
 
Description

Description

  • Synthetic leather palm ensures dexterity and a high level of protection
  • Pre-curved, anatomic design offers a close and comfortable fit
  • Mesh back adds comfort, while also helping to keep the hands cool
  • Elastic wrist provides a secure fit
  • Carabiner loops allow for convenient clipping onto harness gear when not in use
Specifications

Specifications

Mesh
Belay Systems, Power tool Operation, Main Line Hauling, and Repelling
Synthetic Leather
Black
Elasticated
25030
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit