CMC Rescue Riggers Gloves

Perfect blend of dexterity and durability

Manufacturer: CMC RESCUE 250302

 View more versions of this product

Catalog No. 19-100-334

  Check Availability
Add to Cart

    Specifications

    For Use With (Application) to tie knots and work with hardware, set up a system or to rappel
    Size Small
    Material Synthetic Leather
    Color Black

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit