Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CMC Rescue™ Stainless-Steel Rescue Litters

With StratLoad™ attachment points

Manufacturer:  CMC Rescue™ 726100

Catalog No. 19-808-619


Add to cart

Description

Description

  • Full 1 in. dia. top rail makes litter easy to handle when carried for extended periods
  • Durathene™ netting eliminates cuts and scrapes caused by broken wires
  • Molded back support with high density polyethylene for greater durability
  • Steep-sided design allows narrower litter with same width at bed accepting standard-sized backboards
  • Narrow width and rounded ends provides greater maneuverability in tight locations
  • With four quick-attachment litter straps
StartLoad™ Attachment Points
  • Allows any size carabiner to be used to attach lifting bridle while keeping both carabiners and lifting bridle on the inside of litter's top rail
  • Protects from abrasion along cliff faces and building walls
  • Prevents cross gate and side loading of carabiners while orienting direction of pull along spine of carabiner
  • 0.25 in. (6mm) thick with fullround radius on inside edge, allowing attachment protected by top railing
  • Standard on stainless-steel and titanium litters

Certifications

CE marked

Specifications

Specifications

31 lb.
23 in.
Rescue litter
CE
7.25 in.
StratLoad attachment points, full 1 in. dia. top rails makes litter easier to handle, Durathene netting
211cm
14kg
58cm
Safety
83 x 23 x 7.25 in. (211 x 58 x 18.5cm)
18.5cm
83 in.
Stainless Steel
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit