CMC Rescue Training Manuals

Provide rescuers with a reliable and knowledgeable source of training

Manufacturer: CMC RESCUE 993232

 View more versions of this product

Catalog No. 19-808-638

  Check Availability
Add to Cart

    Specifications

    Product Type Rope Rescue Field Guide
    Category Safety and Training Materials
    For Use With (Application) Provides rescuer with a reliable and knowledgeable source of training
    Format Manual

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit