Learn More
CMC Rescue™ Ultra-Pro Edge Protectors
Feature super-slick, high-performance material that protects your ropes from sharp or rough edges and also greatly reduces friction
Manufacturer:  CMC Rescue™ 294044
 View more versions of this product
Catalog No. 19-808-454
Add to cart
Description
Description
  • Offer lighter weight and easier transport than edge rollers or similar devices
  • Have a lower profile to avoid the tipping often encountered with taller devices
  • Conform to the edge and prevent sharp bends in rope
  • Feature carabiner holes in all four corners to allow the Ultra-Pro to be tied off for safety or linked together for extended protection
  • Ultra-Pro 2 accepts two ropes; Ultra-Pro 4 accepts four ropes
Specifications
Specifications
15.6 x 10 in. (41 x 25.4cm)
Edge Protector
Four rope
41cm
25.4cm
425g
Safety
White
15.6 in.
10 in.
15 oz.