Learn More
CMC Rescue™ Steel Carabiners
Manufacturer:  CMC Rescue™ 324001
 View more versions of this product
Catalog No. 19-808-502
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Gates and sleeves constructed of steel, or stainless steel for higher strength, smoother operation, and better corrosion resistance.
·     Gate opening on the large “D” fits over the railings of most stretchers
·     Gate opening on the extra-large “D” fits the rungs on most fire service ladders and stretchers that require a larger carabiner
·     Sleeves lock on gate to prevent jamming or loosening from vibration
Brite finish.
Certifications
UL listed, 19-808-498 and 19-808-500 UL Classified to NFPA 1983, General Use.
Specifications
Specifications
Safety