Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CMIP Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549287

Catalog No. PA549287

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Predicted to react with mouse and xenopus based on sequence homology.

Plays a role in T-cell signaling pathway. Isoform 2 may play a role in T-helper 2 (Th2) signaling pathway and seems to represent the first proximal signaling protein that links T-cell receptor-mediated signal to the activation of c-Maf Th2 specific factor.
Specifications

Specifications

CMIP
Polyclonal
PBS with 0.09% sodium azide
Q8IY22
C-Maf-inducing protein, c-Mip, Truncated c-Maf-inducing protein, Tc-Mip, CMIP, KIAA1694, TCMIP
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
80790
Western Blot
Unconjugated
CMIP
Liquid
CMIP
KLH conjugated synthetic peptide between 735-764 amino acids from the C-terminal region of human CMIP.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit