Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CMKLR1 Rat anti-Human, FITC, Clone: BZ332, Invitrogen™

Rat Monoclonal Antibody

Manufacturer:  Invitrogen MA528098

Catalog No. PIMA528098


Add to cart

Description

Description

Receptor for the chemoattractant adipokine chemerin/RARRES2 and for the omega-3 fatty acid derived molecule resolvin E1. Interaction with RARRES2 induces activation of intracellular signaling molecules, such as SKY, MAPK1/3 (ERK1/2), MAPK14/P38MAPK and PI3K leading to multifunctional effects, like, reduction of immune responses, enhancing of adipogenesis and angionesis. Resolvin E1 down-regulates cytokine production in macrophages by reducing the activation of MAPK1/3 (ERK1/2) and NF-kappa-B. Positively regulates adipogenesis and adipocyte metabolism. Acts as a coreceptor for several SIV strains (SIVMAC316, SIVMAC239, SIVMACL7E-FR and SIVSM62A), as well as a primary HIV-1 strain (92UG024-2). {ECO:0000269; PubMed:15728234, ECO:0000269; PubMed:15753205, ECO:0000269; PubMed:20044979, ECO:0000269; PubMed:9603476}.
Specifications

Specifications

CMKLR1
Monoclonal
1 mg/mL
PBS with 1% BSA and 0.09% sodium azide
Q99788
CMKLR1
Peptide corresponding to an epitope within the N-terminal region of human CMKLR1
Protein G
RUO
Antibody
Monoclonal
Human
Flow Cytometry
BZ332
FITC
CMKLR1
Liquid
Rat
IgG2a
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark.
Primary
1240
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit