Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CMPK1 Mouse anti-Human, Clone: OTI1H8, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA526124

Catalog No. PIMA526124


Add to cart

Description

Description

UCK1 (Uridine-cytidine kinase 1) phosphorylates uridine and cytidine to uridine monophosphate and cytidine monophosphate. This enzyme does not phosphorylate deoxyribonucleosides or purine ribonucleosides. Of note, UCK1 is able to use either ATP or GTP as a phosphate donor. UCK1 also possesses the ability to phosphorylate a number of cytidine and uridine nucleoside analogs such as 6-azauridine, 5-fluorouridine, 4-thiouridine, 5-bromouridine, N(4)-acetylcytidine, N(4)-benzoylcytidine, 5-fluorocytidine, 2-thiocytidine, 5-methylcytidine, and N(4)-anisoylcytidine.
Specifications

Specifications

CMPK1
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
P30085
CMPK1, CMK, CMPK, UCK, UMK, UMPK, UMP-CMP kinase, Cytidine monophosphate kinase, Cytidylate kinase, Deoxycytidylate kinase, Uridine monophosphate kinase, Uridine monophosphate/cytidine monophosphate kinase, UMP/CMP kinase, UMP/CMPK
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
51727
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
OTI1H8
Unconjugated
CMPK1
Liquid
CMPK1
Full length human recombit protein of CMPK1 produced in HEK293T cell
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit