Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CMTM1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA553554
Description
Immunogen sequence: TKMRTNLKRF LGVEVERKLS PAKDAY Highest antigen sequence identity to the following orthologs: Mouse - 38%, Rat - 46%.
Chemokines are members of a superfamily of small inducible, secreted, pro-inflammatory cytokines. Members of the chemokine-like factor superfamily (CKLFSF) share similarities with both chemokine and transmembrane 4 superfamilies of signaling molecules. As a member of the CKLFSF family of proteins, CMTM1 (CKLF-like MARVEL transmembrane domain-containing protein 1), also known as CKLFSF1 (Chemokine-like factor superfamily member 1), is a 169 amino acid multi-pass membrane protein that is highly expressed in testis, therefore probably playing a role in spermatogenesis or testicular development. CMTM1 contains a CC-chemokine motif and four transmembrane segments. The gene encoding CMTM1 resides in a tight gene cluster on human chromosome 16 with CKLF, CMTM2, CMTM3 and CMTM4. There are at least sixteen isoforms of CMTM1 that are produced as a result of alternative splicing events.
Specifications
CMTM1 | |
Polyclonal | |
Unconjugated | |
CMTM1 | |
chemokine-like factor super family 1; chemokine-like factor superfamily 1; chemokine-like factor superfamily member 1; chemokine-like factor-like protein CKLFH1; CHLFH1a; CKLF like MARVEL transmembrane domain containing 1; CKLFH; CKLFH1; CKLF-like MARVEL transmembrane domain containing 1; CKLF-like MARVEL transmembrane domain-containing protein 1; CKLFSF1; CMTM1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
113540 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8IZ96 | |
CMTM1 | |
Recombinant protein corresponding to Human CMTM1. Recombinant protein control fragment (Product #RP-90925). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction