Learn More
Abnova™ CMTM5 293T Cell Transient Overexpression Lysate (Denatured)
Human overexpression lysate
Manufacturer:  Abnova™ H00116173T01
Catalog No. 89-939-948
Add to cart
Description
Description
Specifications
Specifications
Western Blot
CMTM5 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 17.27kDa
Transient overexpression cell lysate was tested with anti-CMTM5 antibody (Mfr. No. H00116173-B01) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
293T
NM_138460
pCMV-CMTM5 full-length
100 μL
Human