Learn More
CNBP Mouse anti-Human, Unconjugated, Clone: 1H1D12, Proteintech
Mouse Monoclonal Antibody
Supplier: Proteintech Group Inc 671091IG150UL
This item is not returnable.
View return policy
Description
The CNBP antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CNBP. This antibody recognizes human, mouse, rat antigen. The CNBP antibody has been validated for the following applications: IP, WB, ELISA analysis.
Specifications
CNBP | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human, Rat, Mouse |
Western Blot, Immunoprecipitation, ELISA | |
1H1D12 | |
Unconjugated | |
150 μL | |
7555 | |
IgG2a |