Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CNDP2 Mouse anti-Human, Clone: OTI8A2, liquid, TrueMAB™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  OriGeneSupplier Diversity Partner TA503168

Catalog No. 50-167-4129


Add to cart

Description

Description

CNDP2 (tissue carnosinase and peptidase A), is a nonspecific dipeptidase rather than a selective carnosinase.
Specifications

Specifications

CNDP2
Monoclonal
0.5 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q96KP4
CN2, CPGL, HEL-S-13, HsT2298, PEPA
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
55748
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
OTI8A2
Unconjugated
CNDP2
Liquid
CNDP2
Full length human recombit protein of human CNDP2 produced in HEK293T cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit