Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CNGA3 Polyclonal Antibody
CNGA3 Polyclonal Antibody
Supplier: Thermo Scientific PA583962
Description
CNGA3 Polyclonal Antibody for IHC (P)

Specifications
CNGA3 | |
Polyclonal | |
Unconjugated | |
CNGA3 | |
ACHM2; alpha subunit of cone photoreceptor CNG-channel; CCNC1; CCNCa; CCNCalpha; CNCG3; cng channel; CNG channel 1; CNG channel alpha-3; CNG1; CNG-1; CNG3; CNG-3; CNGA3; CNGalpha2; cone photoreceptor cGMP-gated channel alpha subunit; cone photoreceptor cGMP-gated channel subunit alpha; cyclic nucleotide gated channel alpha 3; cyclic nucleotide-gated cation channel alpha-3; Cyclic nucleotide-gated channel alpha-3; cyclic nucleotide-gated channel cone photoreceptor subunit alpha | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
1261 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q16281 | |
CNGA3 | |
Recombinant protein corresponding to Human CNGA3. Recombinant protein control fragment (Product #RP-108383). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction