Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CNGA4 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA548284
Description
Second messenger, cAMP, causes the opening of cation- selective cyclic nucleotide-gated (CNG) channels and depolarization of the neuron (olfactory sensory neurons, OSNs). CNGA4 is the modulatory subunit of this channel which is known to play a central role in the transduction of odorant signals and subsequent adaptation. By accelerating the calcium-mediated negative feedback in olfactory signaling it allows rapid adaptation to odor stimulation and extends its range of odor detection.Specifications
CNGA4 | |
Polyclonal | |
PBS with 0.09% sodium azide | |
Q8IV77, Q3UW12 | |
CNGA4 | |
KLH conjugated synthetic peptide between 131-160 amino acids from the N-terminal region of human CNGA4. | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
CNGA4 | |
Cyclic nucleotide-gated cation channel alpha-4, Cyclic nucleotide-gated channel alpha-4, CNG channel alpha-4, CNG-4, CNG4, CNGA4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
1262, 233649 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction