Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CNGB3 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA548428
Description
Visual signal transduction is mediated by a G-protein coupled cascade using cGMP as second messenger. This protein can be activated by cGMP which leads to an opening of the cation channel and thereby causing a depolarization of rod photoreceptors. Induced a flickering channel gating, weakened the outward rectification in the presence of extracellular calcium, increased sensitivity for L-cis diltiazem and enhanced the cAMP efficiency of the channel when coexpressed with CNGA3. Essential for the generation of light-evoked electrical responses in the red-, green- and blue sensitive cones.Specifications
CNGB3 | |
Polyclonal | |
PBS with 0.09% sodium azide | |
Q9NQW8 | |
CNGB3 | |
KLH conjugated synthetic peptide between 26-55 amino acids from the N-terminal region of human CNGB3. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
CNGB3 | |
Cyclic nucleotide-gated cation channel beta-3, Cone photoreceptor cGMP-gated channel subunit beta, Cyclic nucleotide-gated cation channel modulatory subunit, Cyclic nucleotide-gated channel beta-3, CNG channel beta-3, CNGB3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
54714 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction