Learn More
Cnidaria Products for Science Education
Use these animal specimens to compare the three classes of Cnidaria.
Specifications
Quantity 10 Pack
View More Specs
Products 6
Catalog Number Length (English) Preservative Product Type Price Quantity & Availability  
Catalog Number Length (English) Preservative Product Type Price Quantity & Availability  
S1001S Products for Science Education
View Documents
Alcohol Hydra
Pack of 10 for $8.68
N/A
Due to product restrictions, please to purchase or view availability for this product.
S1282S Products for Science Education
View Documents
Bio-Fresh™ Jellyfish, Portuguese Man-O-War
Pack for $21.00
N/A
Due to product restrictions, please to purchase or view availability for this product.
S1360S Products for Science Education
View Documents
4 to 6 in. Bio-Fresh™ Common Jellyfish, Aurelia
Pack of 10 for $58.46
N/A
Due to product restrictions, please to purchase or view availability for this product.
S1499S Products for Science Education
View Documents
1.5 to 2 in. Bio-Fresh™ Sea Anemone
Pack of 10 for $41.54
N/A
Due to product restrictions, please to purchase or view availability for this product.
S2502 Products for Science Education
View Documents
3 to 4 in. Bio-Fresh™ Common Jellyfish, Aurelia
Pack of 10 for $42.35
N/A
Due to product restrictions, please to purchase or view availability for this product.
S1500S Products for Science Education
View Documents
2+ in. Bio-Fresh™ Sea Anemone This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Due to product restrictions, please to purchase or view availability for this product.
N/A
Description
Description
Hydra
  • Common freshwater hydrozoans that can be encountered on field trips
  • Students can use these specimens for anatomical studies and for familiarity before going out on a field trip to an aquatic environment
  • In alcohol
Sea Anemone (Metridium)
  • Most common Anthozoan studied
  • Only well-expanded specimens are offered
  • Available in three sizes in Fisher Science Education™ Bio-Fresh™ solution
Portuguese Man-O-War (Physalia)
  • Physalia dramatically demonstrates the various groups of specialized cell members within the hydroid colony
  • Students especially enjoy these spectacular animal specimens
Common Moon Jellyfish (Aurelia)
  • Aurelia are ciliary feeders in the scyphozoan class
  • As they sink, plankton becomes entrapped on the mucus of the subumbrellar surface
  • From here, flagella sweep the food into the Aurelia's mouth
  • In Fisher Science Education Bio-Fresh solution
Specifications
Specifications
10 Pack