Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CNNM1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP31549720UL
Description
CNNM1 Polyclonal antibody specifically detects CNNM1 in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
CNNM1 | |
Polyclonal | |
PBS, 50% glycerol, pH7.3 | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunofluorescence | |
Unconjugated | |
ACDP1cyclin-M1, ancient conserved domain protein 1, Ancient conserved domain-containing protein 1, cyclin M1, Cyclin-M1, FLJ31632, metal transporter CNNM1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 692-951 of human CNNM1 (NP_065081.2). | |
20 μg | |
Cell Biology, Cell Cycle and Replication | |
26507 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction