Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CNNM4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA595885
Description
Immunogen sequence: VRALVDLQYI KITRQQYQNG LLASRMENSP QFPIDGCTTH MENLAEKSEL PVVDETTTLL NERNSLLHKA SHENAI.
This gene encodes a member of the ancient conserved domain containing protein family. Members of this protein family contain a cyclin box motif and have structural similarity to the cyclins. The encoded protein may play a role in metal ion transport. Mutations in this gene are associated with Jalili syndrome which consists of cone-rod dystrophy and amelogenesis imperfecta.
Specifications
CNNM4 | |
Polyclonal | |
Unconjugated | |
CNNM4 | |
5430430O18Rik; Acdp4; ancient conserved domain protein 4; ancient conserved domain-containing protein 4; Cnnm4; cyclin and CBS domain divalent metal cation transport mediator 4; cyclin M4; cyclin-M4; Kiaa1592; mACDP4; metal transporter CNNM4 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
26504, 363216, 94220 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
1.6 mg/mL | |
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
P0C588, Q69ZF7, Q6P4Q7 | |
CNNM4 | |
A synthetic peptide corresponding to a sequence within amino acids 700-775 of human CNNM4 (NP_0645693). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction