Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CNOT2 Mouse anti-Human, Janelia Fluor™ 646, Clone: OTI5A12, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP271202JF646

Catalog No. N271202J646


Add to cart

Description

Description

CNOT2 Monoclonal antibody specifically detects CNOT2 in Human samples. It is validated for Immunohistochemistry.
Specifications

Specifications

CNOT2
OTI5A12
Immunohistochemistry
Affinity Purified, Conjugated
CNOT2
Full length recombinant protein of human CNOT2 (NP_055330) produced in E.coli.
Immunogen affinity purified
Store at 4°C in the dark.
Monoclonal
Human
Immunohistochemistry
Janelia Fluor 646
50mM Sodium Borate with 0.05% Sodium Azide
CCR4-associated factor 2, CCR4-NOT transcription complex, subunit 2, CDC36FLJ26456, negative regulator of transcription 2, NOT2CCR4-NOT transcription complex subunit 2, NOT2H
Mouse
IgG2b
0.1 mL
Primary
4848.0
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit