Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CNOT4 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$206.50 - $426.00

Specifications

Antigen CNOT4
Concentration 0.5 mg/mL
Dilution Western Blot 0.05-0.1 ug/ml, Immunohistochemistry-Paraffin 10 ug/ml
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP224527SS
SDP
View Documents
Novus Biologicals
NBP224527SS
0.025 mg
Each for $206.50
Only null left
Add to Cart
 
NBP224527
SDP
View Documents
Novus Biologicals
NBP224527
0.1 mg
Each for $426.00
Only null left
Add to Cart
 
Description

Description

CNOT4 Polyclonal specifically detects CNOT4 in Human, Mouse, Rat, Bovine, Chicken, Xenopus samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

CNOT4
Western Blot 0.05-0.1 ug/ml, Immunohistochemistry-Paraffin 10 ug/ml
Polyclonal
Rabbit
Human, Mouse, Rat, Bovine, Chicken, Xenopus species
CCR4-associated factor 4, CCR4-NOT transcription complex, subunit 4, E3 ubiquitin-protein ligase CNOT4, EC 6.3.2.-, NOT4yeast) homolog
CNOT4
IgG
Affinity Purified
The amino acid sequence used as immunogen for the antibody is 100% homologous in human, rhesus monkey, mouse, rat, dog and chimpanzee, 93% homologous in opossum, 86% homologous in chicken and 73% homologous in xenopus.
0.5 mg/mL
Western Blot, Immunohistochemistry (Paraffin)
Unconjugated
RUO
0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide
4850
A portion of amino acids 50-100 of human CNOT4 was used as the immunogen for the antibody.
Primary
Store at -20C. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.