Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CNOT4 Antibody (OTI3D11), Janelia Fluor™ 549, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271203JF549
Description
CNOT4 Monoclonal specifically detects CNOT4 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry.Specifications
CNOT4 | |
Monoclonal | |
Janelia Fluor 549 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
CNOT4 | |
Recombinant protein fragment corresponding to amino acids 190-455 of human CNOT4 (NP_037448) produced in E.coli. | |
0.1 mL | |
4850.0 | |
Store at 4°C in the dark. |
Western Blot, Flow Cytometry | |
OTI3D11 | |
Western Blot, Flow Cytometry | |
CCR4-associated factor 4, CCR4-NOT transcription complex, subunit 4, E3 ubiquitin-protein ligase CNOT4, EC 6.3.2.-, NOT4yeast) homolog | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction