Learn More
Abnova™ CNP 293T Cell Transient Overexpression Lysate (Denatured)
Human overexpression lysate
Manufacturer:  Abnova™ H00001267T01
Catalog No. 89-940-019
Add to cart
Description
Description
Specifications
Specifications
Western Blot
CNP 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 46.42kDa
Transient overexpression cell lysate was tested with anti-CNP antibody (Mfr. No. H00001267-B01) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
293T
NM_033133.4
pCMV-CNP full-length
100 μL
Human