Learn More
Aviva Systems Biology Corp CNP ANTIBODY 50UG/50UL DFS Item
Manufacturer:  Aviva Systems Biology Corp ARP33879_P050
This is a rabbit polyclonal antibody against CNP. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human CNP. Alternate names: CNP1ApplicationIHC, WB. Species reactivity: Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-55185