Learn More
CNQX disodium salt, Tocris
Potent AMPA/kainate antagonist; More water soluble form of CNQX
Manufacturer:  R&D Systems 1045/50
Quantity 50mg
 View more versions of this product
Catalog No. 10-455-0
  Check Availability
Add to cart
Specifications Description & Specifications
Chemical Identifiers
>99%
276.12
CNQX disodium salt
C9H2N4O4Na2
6-Cyano-7-nitroquinoxaline-2, 3-dione disodium
Specifications
50mg
>99%
276.12
CNQX disodium salt
479347-85-8
C9H2N4O4Na2
6-Cyano-7-nitroquinoxaline-2, 3-dione disodium
Safety and Handling
Recommended Storage : Desiccate at Room Temperature