Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CNQX, Tocris Bioscience™

Catalog Number 019010
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
115066-14-3
C9H4N4O4
232.155
RPXVIAFEQBNEAX-UHFFFAOYSA-N
cnqx, 6-cyano-7-nitroquinoxaline-2,3-dione, unii-6ote87sccw, 7-nitro-2,3-dioxo-1,2,3,4-tetrahydroquinoxaline-6-carbonitrile, 6ote87sccw, 3h cnqx, 6-cyano-2,3-dihydroxy-7-nitroquinoxaline, 1,4-dihydro-2,3-dihydroxy-7-nitro-6-quinoxalinecarbonitrile, 6-quinoxalinecarbonitrile, 1,2,3,4-tetrahydro-7-nitro-2,3-dioxo, 2,3-dihydroxy-7-nitro-6-quinoxalinecarbonitrile
3721046
CHEBI:34468
7-nitro-2,3-dioxo-1,4-dihydroquinoxaline-6-carbonitrile
C1=C(C(=CC2=C1NC(=O)C(=O)N2)[N+](=O)[O-])C#N
This item is not returnable. View return policy
115066-14-3
C9H4N4O4
232.155
RPXVIAFEQBNEAX-UHFFFAOYSA-N
cnqx, 6-cyano-7-nitroquinoxaline-2,3-dione, unii-6ote87sccw, 7-nitro-2,3-dioxo-1,2,3,4-tetrahydroquinoxaline-6-carbonitrile, 6ote87sccw, 3h cnqx, 6-cyano-2,3-dihydroxy-7-nitroquinoxaline, 1,4-dihydro-2,3-dihydroxy-7-nitro-6-quinoxalinecarbonitrile, 6-quinoxalinecarbonitrile, 1,2,3,4-tetrahydro-7-nitro-2,3-dioxo, 2,3-dihydroxy-7-nitro-6-quinoxalinecarbonitrile
3721046
CHEBI:34468
7-nitro-2,3-dioxo-1,4-dihydroquinoxaline-6-carbonitrile
C1=C(C(=CC2=C1NC(=O)C(=O)N2)[N+](=O)[O-])C#N

Potent and selective non-NMDA iGluR antagonist

CNQX is an AMPA and kainate receptor antagonist (IC50 values are 0.3 μM and 1.5 μM for AMPA and kainate receptors, respectively). CNQX is also an antagonist at the glycine modulatory site on the NMDA receptor complex (IC50 = 25 μM). CNQX can be used to isolate GABAA receptor-mediated spontaneous inhibitory postsynaptic currents and antagonizes non-NMDA receptor-mediated responses in cultured cerebellar granule cells. CNQX shows neuroprotective effects in models of ischemia and inhibits seizure-like activity in hippocampal neurons.

CNQX Disodium Salt (Cat. No. 1045) also available.

Quantity 10 mg
Formula Weight 232.16
Percent Purity >99%
Chemical Name or Material CNQX
Recommended Storage Store at Room Temperature
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit