Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CNTF Mouse anti-Human, Mouse, Rat, Primate, Rabbit, Clone: 24929, Novus Biologicals™
Mouse Monoclonal Antibody
$212.56 - $804.00
Specifications
Antigen | CNTF |
---|---|
Clone | 24929 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:100-1:2000, Immunohistochemistry 10-20 ug/ml |
Applications | Western Blot, Immunohistochemistry |
Description
CNTF Monoclonal specifically detects CNTF in Human, Mouse, Rat, Primate, Rabbit samples. It is validated for Western Blot, Immunohistochemistry.Specifications
CNTF | |
1 mg/mL | |
Western Blot, Immunohistochemistry | |
Unconjugated | |
RUO | |
Human, Mouse, Rat, Primate, Rabbit | |
ciliary neurotrophic factor, HCNTF | |
CNTF | |
IgG1 | |
Affinity Purified | |
Ciliary neurotrophic factor. Detects a 22 kD band in Western Blots of rat sciatic nerve extract. |
24929 | |
Western Blot 1:100-1:2000, Immunohistochemistry 10-20 ug/ml | |
Monoclonal | |
Mouse | |
Neuroscience | |
P26441 | |
1270 | |
Purified ciliary neurotrophic factor from rat sciatic nerve. | |
Primary | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
For Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title