Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CNTFR Mouse anti-Human, Alexa Fluor™ 750, Clone: MM0190-10L29, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP212196AF750

Catalog No. NP212196A70


Add to cart

Description

Description

CNTFR Monoclonal antibody specifically detects CNTFR in Human samples. It is validated for Western Blot,Immunohistochemistry,Immunohistochemistry (Paraffin).
Specifications

Specifications

CNTFR
MM0190-10L29
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
ciliary neurotrophic factor receptor, ciliary neurotrophic factor receptor alpha, ciliary neurotrophic factor receptor subunit alpha, CNTF receptor subunit alpha, CNTFR alpha, CNTFR-alpha, MGC1774
Mouse
IgG1
Store at 4°C in the dark.
Monoclonal
Human
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Alexa Fluor™ 750
50 mM sodium borate with 0.05% sodium azide
CNTFR
Human CNTFR
0.1 mL
Primary
1271
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit