Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CNTLN Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548521

Catalog No. PA548521

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Required for centrosome cohesion and recruitment of CEP68 to centrosomes.
Specifications

Specifications

CNTLN
Polyclonal
PBS with 0.09% sodium azide
Q9NXG0
Centlein, Centrosomal protein, CNTLN, C9orf101, C9orf39
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
54875
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
CNTLN
Liquid
CNTLN
KLH conjugated synthetic peptide between 199-227 amino acids from the N-terminal region of human CNTLN.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit