Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CNTNAP4 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$436.00
Specifications
Antigen | CNTNAP4 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP159223
|
Novus Biologicals
NBP159223 |
100 μL |
Each of 1 for $436.00
|
|
Description
CNTNAP4 Polyclonal specifically detects CNTNAP4 in Human samples. It is validated for Western Blot.Specifications
CNTNAP4 | |
Polyclonal | |
Rabbit | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
CASPR4KIAA1763cell recognition protein CASPR4, Cell recognition molecule Caspr4, contactin associated protein-like 4, contactin-associated protein-like 4 | |
CNTNAP4 | |
IgG | |
Affinity Purified |
Western Blot | |
Unconjugated | |
RUO | |
Q9C0A0 | |
85445 | |
Synthetic peptides corresponding to CNTNAP4(contactin associated protein-like 4) The peptide sequence was selected from the N terminal of CNTNAP4. Peptide sequence KLPSTSTLVNLTLGSLLDDQHWHSVLIQRLGKQVNFTVDEHRHHFHARGE. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title