Learn More
Co 102862, Tocris
State-dependent Na+ channel blocker; anticonvulsant
Manufacturer:  R&D Systems 2642/50
Quantity 50mg
 View more versions of this product
Catalog No. 26-425-0
  Check Availability
Add to cart
Specifications Description & Specifications
Chemical Identifiers
>99%
273.26
Co 102862
C14H12FN3O2
2-[[4-(4-Fluorophenoxy)phenyl]methylene]hydrazinecarboxamide
Specifications
50mg
>99%
273.26
Co 102862
181144-66-1
C14H12FN3O2
2-[[4-(4-Fluorophenoxy)phenyl]methylene]hydrazinecarboxamide
Safety and Handling
Recommended Storage : Store at Room Temperature