Learn More
UbpbioSupplier Diversity Partner CO-NTA AGAROSE, 100ML DFS Item
Manufacturer:  UbpbioSupplier Diversity Partner P3030100
Protein Purification Reagents – Co-NTA Resin (UBPBio catalog# P3030-5, P3030-25, P3030-100) – Co-NTA XPure Agarose Resin is intended for preparative purification of histidine-tagged recombinant proteins from all prokaryotic and eukarotic expression systems. Co-NTA XPure Agarose Resin consists of highly cross-linked 6% agarose with an immobilized chelating group. The talon ligand is a tetra-dentate chelator charged with cobalt. Co-NTA XPure Agarose Resin offers enhanced selectivity for histidine-tagged proteins compared to nickel-charged medium.
Catalog No. 50-112-6951