Thermo Scientific™ CO2 and LN2 Backup Systems for Ultra-Low Temperature Freezers Available on GSA/VA Contract for Federal Government customers only.
Ensure optimal security for critical samples by using Thermo Scientific™ CO2 and LN2 Backup Systems for Ultra-Low Temperature Freezers.
Manufacturer: Thermo Scientific™ 1950445
 View more versions of this product
Catalog No. 1950445
  Check Availability
Add to Cart
    Specifications
    For Use With (Equipment) Forma™ 900, 7000 and 8600 Series
    Type CO2 Backup System,Factory-Installed
    Voltage N.A.
    Temperature Maintains temperature down to minus70°C
    Description CO2 Backup System
    For Use With Forma 900, 7000 and 8600 Series; HFU B and HFC Series; Revco CxF Series; TSC Series
    Item Description 1950445
    Ensure optimal security for critical samples by using Thermo Scientific™ CO2 and LN2 Backup Systems for Ultra-Low Temperature Freezers, for use with Thermo Scientific™ Forma™, Thermo Scientific™ HERAfreeze™, Thermo Scientific™ Revco™ and TS-Series freezers.
    Features:
    • LN2 backup system maintains temperature down to -80°C with liquid nitrogen
    • CO2 backup system maintains temperature down to -7° or -70°C (model dependant) with CO2
    Ordering Information:
    Available as field- or factory-installed. Models for UxF300/88300/HFU300T/TSU300 Series are standalone. Field-installed units must be installed by qualified professionals.
  • Description & Specifications
Related Products
Product Content Feedback