Learn More
Thermo Scientific™ CO2 and LN2 Backup Systems for Ultra-Low Temperature Freezers
Ensure optimal security for critical samples by using Thermo Scientific™ CO2 and LN2 Backup Systems for Ultra-Low Temperature Freezers.
Manufacturer:  Thermo Scientific™ 1950534
 View more versions of this product
Catalog No. 1950534
  Check Availability
Add to cart
Specifications
Description
Ensure optimal security for critical samples by using Thermo Scientific™ CO2 and LN2 Backup Systems for Ultra-Low Temperature Freezers, for use with Thermo Scientific™ Forma™, Thermo Scientific™ HERAfreeze™, Thermo Scientific™ Revco™ and TS-Series freezers.
Features:
  • LN2 backup system maintains temperature down to -80°C with liquid nitrogen
  • CO2 backup system maintains temperature down to -7° or -70°C (model dependant) with CO2
Ordering Information:
Available as field- or factory-installed. Models for UxF300/88300/HFU300T/TSU300 Series are standalone. Field-installed units must be installed by qualified professionals.
Description & Specifications
Specifications
CO2 Backup System
CO2 Backup System,Field-Installed
Maintains temperature down to minus70°C
TSE and TSD ULT Series; Revco ExF, DxF, CxF Series
N.A.
Revco™ ExF, DxF, CxF Series