Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ COA4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA559076
Description
Immunogen sequence: MSTSVPQGHT WTQRVKKDDE EEDPLDQLIS RSGCAASHFA VQECMAQHQD WRQCQPQVQA FKDCMSEQQA RRQEE Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 89%.
Putative COX assembly factor.
Specifications
COA4 | |
Polyclonal | |
Unconjugated | |
COA4 | |
5330414O08Rik; AI115477; AW047259; CHCHD8; CMC3; Coa4; coiled-coil-helix-coiled-coil-helix domain containing 8; coiled-coil-helix-coiled-coil-helix domain-containing protein 8; cytochrome c oxidase assembly factor 4; cytochrome c oxidase assembly factor 4 homolog; cytochrome c oxidase assembly factor 4 homolog, mitochondrial; E2ig2; E2-induced gene 2 protein; RGD1562228 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
51287 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.7 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9NYJ1 | |
COA4 | |
Recombinant protein corresponding to Human COA4. Recombinant protein control fragment (Product #RP-97518). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction