Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Coactosin-like Protein 1/CotL1 Mouse anti-Human, Alexa Fluor™ 350, Clone: 1D6, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP231203AF350

Catalog No. NP231203A30


Add to cart

Description

Description

Coactosin-like Protein 1/CotL1 Monoclonal antibody specifically detects Coactosin-like Protein 1/CotL1 in Human samples. It is validated for Western Blot,ELISA.
Specifications

Specifications

Coactosin-like Protein 1/CotL1
1D6
Western Blot, ELISA
CLPFLJ43657, coactosin-like 1 (Dictyostelium), coactosin-like protein, MGC19733
Mouse
IgG2b Kappa
Store at 4°C in the dark.
Monoclonal
Human
ELISA, Western Blot
Alexa Fluor™ 350
50 mM sodium borate with 0.05% sodium azide
COTL1
Recombinant human COTL1 (1-142aa) purified from E. coli
0.1 mL
Primary
23406
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit