Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Coactosin-like Protein 1/CotL1 Antibody (1D6), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP231203AF532
Description
Coactosin-like Protein 1/CotL1 Monoclonal specifically detects Coactosin-like Protein 1/CotL1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.Specifications
Coactosin-like Protein 1/CotL1 | |
Monoclonal | |
Alexa Fluor 532 | |
50 mM sodium borate with 0.05% sodium azide | |
COTL1 | |
Recombinant human COTL1 (1-142aa) purified from E. coli | |
Cytokine Research, Signal Transduction | |
23406 | |
Store at 4°C in the dark. |
Western Blot, Flow Cytometry, ELISA | |
1D6 | |
Western Blot, Flow Cytometry, ELISA | |
CLPFLJ43657, coactosin-like 1 (Dictyostelium), coactosin-like protein, MGC19733 | |
Mouse | |
0.1 mL | |
Primary | |
Human | |
IgG2b κ |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction