Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Coactosin-like Protein 1/CotL1 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP15485120UL

 View more versions of this product

Catalog No. NBP1548520


Add to cart

Description

Description

Coactosin-like Protein 1/CotL1 Polyclonal antibody specifically detects Antigen in Human samples. It is validated for Western Blotting.
Specifications

Specifications

Coactosin-like Protein 1/CotL1
Unconjugated
PBS and 2% Sucrose with 0.09% Sodium Azide
Affinity Purified
COTL1
Synthetic peptides corresponding to COTL1(coactosin-like 1 (Dictyostelium)) The peptide sequence was selected from the N terminal of COTL1. Peptide sequence MATKIDKEACRAAYNLVRDDGSAVIWVTFKYDGSTIVPGEQGAEYQHFIQ.
16 kDa
20ul
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Human
Western Blot
Western Blot 1:100-1:2000
Q14019
CLPFLJ43657, coactosin-like 1 (Dictyostelium), coactosin-like protein, MGC19733
Rabbit
IgG
Immunogen affinity purified
RUO
Primary
23406
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit