Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Coagulation Factor II/Thrombin Mouse anti-Human, Alexa Fluor™ 594, Clone: 22, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP241381AF594

Catalog No. NP241381A94


Add to cart

Description

Description

Coagulation Factor II/Thrombin Monoclonal antibody specifically detects Coagulation Factor II/Thrombin in Human samples. It is validated for ELISA,Immunohistochemistry.
Specifications

Specifications

Coagulation Factor II/Thrombin
22
ELISA, Immunohistochemistry, Sandwich ELISA
Coagulation Factor II, coagulation factor II (thrombin) receptor-like 2, Coagulation factor II receptor-like 2, Coagulation factor II receptor-like 2 (protease-actovated receptor 3), PAR-3, PAR3proteinase-activated receptor 3, protease-activated receptor 3, proteinase-activated receptor-3, PT, serine protease, Thrombin receptor-like 2
Mouse
IgG1 Kappa
Store at 4°C in the dark.
Monoclonal
Antibody is specific for human Thrombin-antithrombin (TAT) complexes.
ELISA, Immunohistochemistry
Alexa Fluor™ 594
50 mM sodium borate with 0.05% sodium azide
F2
Thrombin-antithrombin (TAT) complexes
0.1 mL
Primary
2147
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit