Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thrombin-antithrombin (TAT) complexes Antibody (22), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP241381B
Description
Thrombin-antithrombin (TAT) complexes Monoclonal antibody specifically detects Thrombin-antithrombin (TAT) complexes in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.Specifications
Thrombin-antithrombin (TAT) complexes | |
Monoclonal | |
Biotin | |
PBS with 0.05% Sodium Azide | |
F2 | |
Thrombin-antithrombin (TAT) complexes | |
0.1 mL | |
Primary | |
Antibody is specific for human Thrombin-antithrombin (TAT) complexes. | |
Store at 4C in the dark. | |
IgG1 κ |
Western Blot, ELISA, Immunohistochemistry | |
22 | |
Western Blot, ELISA, Immunohistochemistry, Sandwich ELISA | |
Coagulation Factor II, coagulation factor II (thrombin) receptor-like 2, Coagulation factor II receptor-like 2, Coagulation factor II receptor-like 2 (protease-actovated receptor 3), PAR-3, PAR3proteinase-activated receptor 3, protease-activated receptor 3, proteinase-activated receptor-3, PT, serine protease, Thrombin receptor-like 2 | |
Mouse | |
Protein A or G purified | |
Apoptosis, Breast Cancer, Cancer, GPCR, Signal Transduction | |
2147 | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction