Learn More
Coagulation Factor II/Thrombin Mouse anti-Human, DyLight 650, Clone: 22, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP241381C
Catalog No. NBP241381C
Add to cart
Description
Description
Coagulation Factor II/Thrombin Monoclonal antibody specifically detects Coagulation Factor II/Thrombin in Human samples. It is validated for Western Blot.
Specifications
Specifications
Coagulation Factor II/Thrombin
22
Western Blot
Purified
F2
Thrombin-antithrombin (TAT) complexes
Protein A or G purified
Apoptosis, Breast Cancer, Cancer, GPCR, Signal Transduction
Primary
2147
Human
Western Blot
DyLight 650
50mM Sodium Borate with 0.05% Sodium Azide
Coagulation Factor II, coagulation factor II (thrombin) receptor-like 2, Coagulation factor II receptor-like 2, Coagulation factor II receptor-like 2 (protease-actovated receptor 3), PAR-3, PAR3proteinase-activated receptor 3, protease-activated receptor 3, proteinase-activated receptor-3, PT, serine protease, Thrombin receptor-like 2
Mouse
IgG1 Kappa
0.1 ml
Store at 4C in the dark.
Monoclonal
Antibody is specific for human Thrombin-antithrombin (TAT) complexes.