Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Coagulation Factor III/Tissue Factor Antibody Pair [HRP]

Polyclonal/Monoclonal Antibody Pair

Manufacturer:  Novus Biologicals™ NBP2796505PLATES

 View more versions of this product

Catalog No. NB010340


Add to cart

Description

Description

This solid phase sandwich ELISA utilizes an antibody specific for Rat Coagulation Factor III/Tissue Factor.
Specifications

Specifications

23.45 pg/mL
Conjugated
2152
Antibody Pairs
23.45-1500 pg/mL
Storage is content dependent.
Rabbit/Mouse
5 Plates
Sandwich ELISA
CD142, CD142 antigen, Coagulation factor III, coagulation factor III (thromboplastin, tissue factor), FLJ17960, TF, TFA, Thromboplastin, tissue factor
HRP
Sandwich ELISA
  • Standard: Each vial contains 80 ng of recombinant Rat Coagulation Factor III/Tissue Factor.
  • Capture Antibody: 0.5 mg/mL of mouse anti-Rat Coagulation Factor III/Tissue Factor monoclonal antibody (in PBS, pH 7.4).
  • Detection Antibody: 0.2 mg/mL of rabbit anti-Coagulation Factor III/Tissue Factor polyclonal antibody conjugated to horseradish-peroxidase (HRP) (in PBS, 50 % HRP-Protector, pH 7.4, store at 4C).
  • RUO
    This solid phase sandwich ELISA utilizes an antibody specific for Rat Coagulation Factor III/Tissue Factor.
    Coagulation Factor III/Tissue Factor
    Cancer
    Rat
    F3
    1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in PBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.5 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 1500 pg/mL is recommended.
    SDS
    Documents
    Provide Content Correction

    We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Cancel Submit