Learn More
Coagulation Factor III/Tissue Factor Mouse anti-Human, Alexa Fluor™ 700, Clone: 323519, R&D Systems™
Mouse Monoclonal Antibody
Manufacturer:  R&D Systems FAB23391N100UG
Catalog No. FB23391N100
Add to cart
This item is not returnable. View return policy
Description
Description
Coagulation Factor III/Tissue Factor Monoclonal antibody specifically detects Coagulation Factor III/Tissue Factor in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
Coagulation Factor III/Tissue Factor
323519
Flow Cytometry 0.25-1 μg/106 cells
P13726
CD142, CD142 antigen, Coagulation factor III, coagulation factor III (thromboplastin, tissue factor), F3, FLJ17960, TF, TFA, Thromboplastin, tissue factor
Mouse
IgG1
100 μg
Primary
2152
Human
Flow Cytometry
Alexa Fluor 700
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Conjugated, Purified, Supernatant
F3
Mouse myeloma cell line NS0-derived recombinant human Coagulation Factor III, Gly34-Glu251, Accession # P13726
Protein A or G purified from hybridoma culture supernatant
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Monoclonal
Detects human Coagulation Factor III/Tissue Factor in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse Coagulation Factor III/Tissue Factor is observed.