Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Coagulation Factor VII Antibody Pair [HRP]

Rabbit Monoclonal Antibody Pair

Manufacturer:  Novus Biologicals™ NBP2794255PLATES

 View more versions of this product

Catalog No. NB009890


Add to cart

Description

Description

This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Mouse Coagulation Factor VII.
Specifications

Specifications

Sandwich ELISA
Antibody Pairs
25 pg/mL
2155
RUO
Coagulation Factor VII
5 Plates
25-1600 pg/mL
coagulation factor VII, coagulation factor VII (serum prothrombin conversion accelerator), EC 3.4.21, EC 3.4.21.21, Eptacog alfa, FVII coagulation protein, Proconvertin, Serum prothrombin conversion accelerator, SPCA
HRP
This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Mouse Coagulation Factor VII.
Sandwich ELISA
  • Detection Antibody: 0.2 mg/mL of rabbit anti-Coagulation Factor VII monoclonal antibody conjugated to horseradish-peroxidase (HRP) (in PBS, 50 % HRP-Protector, pH 7.4, store at 4C).
  • Capture Antibody: 0.2 mg/mL of rabbit anti-Mouse Coagulation Factor VII monoclonal antibody (in PBS, pH 7.4).
  • Standard: Each vial contains 28.5 ng of recombinant Mouse Coagulation Factor VII.
  • Storage is content dependent.
    Rabbit
    Mouse
    F7
    1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in PBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.25 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 1600 pg/mL is recommended.
    SDS
    Documents
    Product Certifications
    Promotions

    Promotions

    Provide Content Correction

    We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Cancel Submit