Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Coagulation Factor VII Mouse anti-Human, Clone: F7/3511, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP30724310UG

 View more versions of this product

Catalog No. NB121959


Add to cart

Description

Description

Coagulation Factor VII Monoclonal specifically detects Coagulation Factor VII in Human samples. It is validated for ELISA, Immunohistochemistry-Paraffin, Protein Array.
Specifications

Specifications

Coagulation Factor VII
F7/3511
Unconjugated
10 mM PBS with 0.05% BSA
coagulation factor VII, coagulation factor VII (serum prothrombin conversion accelerator), EC 3.4.21, EC 3.4.21.21, Eptacog alfa, FVII coagulation protein, Proconvertin, Serum prothrombin conversion accelerator, SPCA
Recombinant fragment (around aa 366-466) of human Coagulation Factor VII protein
Protein A or G purified
RUO
Primary
2155
ELISA, Immunohistochemistry (Paraffin), Peptide Array
0.2 mg/ml
ELISA, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array
Purified
Mouse
IgG2a Kappa
100 μg
Store at 4°C.
Monoclonal
Human
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit