Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Coagulation Factor VII Mouse anti-Human, Clone: F7/3511, Novus Biologicals™
Mouse Monoclonal Antibody
$265.00
Specifications
Antigen | Coagulation Factor VII |
---|---|
Clone | F7/3511 |
Concentration | 0.2 mg/ml |
Dilution | ELISA, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array |
Applications | ELISA, Immunohistochemistry (Paraffin), Peptide Array |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB121960
|
Novus Biologicals
NBP30724320UG |
20 μg |
Each of 1 for $265.00
|
|
|||||
Description
Coagulation Factor VII Monoclonal specifically detects Coagulation Factor VII in Human samples. It is validated for ELISA, Immunohistochemistry-Paraffin, Protein Array.Specifications
Coagulation Factor VII | |
0.2 mg/ml | |
ELISA, Immunohistochemistry (Paraffin), Peptide Array | |
Unconjugated | |
Mouse | |
Human | |
coagulation factor VII, coagulation factor VII (serum prothrombin conversion accelerator), EC 3.4.21, EC 3.4.21.21, Eptacog alfa, FVII coagulation protein, Proconvertin, Serum prothrombin conversion accelerator, SPCA | |
Recombinant fragment (around aa 366-466) of human Coagulation Factor VII protein | |
Primary | |
Store at 4°C. |
F7/3511 | |
ELISA, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
Monoclonal | |
Purified | |
RUO | |
10 mM PBS with 0.05% BSA | |
2155 | |
IgG2a κ | |
Protein A or G purified |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title