Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Coagulation Factor X Antibody Pair [HRP]

Monoclonal Antibody Pair

Manufacturer:  Novus Biologicals™ NBP2793425PLATES

 View more versions of this product

Catalog No. NB009724


Add to cart

Description

Description

This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human Coagulation Factor X.
Specifications

Specifications

Sandwich ELISA
Antibody Pairs
5.47 pg/mL
2159
RUO
Coagulation Factor X
5 Plates
5.47-350 pg/mL
coagulation factor X, EC 3.4.21, EC 3.4.21.6, factor Xa, FX, FXA, prothrombinase, Stuart factor, Stuart-Prower factor
HRP
This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human Coagulation Factor X.
Sandwich ELISA
  • Standard: Each vial contains 30 ng of recombinant Human Coagulation Factor X.
  • Capture Antibody: 1 mg/mL of mouse anti-Human Coagulation Factor X monoclonal antibody (in PBS, pH 7.4).
  • Detection Antibody: 0.2 mg/mL of rabbit anti-Human Coagulation Factor X monoclonal antibody conjugated to horseradish-peroxidase (HRP) (in PBS, 50 % HRP-Protector, pH 7.4, store at 4C).
  • Storage is content dependent.
    Rabbit/Mouse
    Human
    F10
    1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in PBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.5 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 350 pg/mL is recommended.
    SDS
    Documents
    Product Certifications
    Promotions

    Promotions

    Provide Content Correction

    We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Cancel Submit