Learn More
Coagulation Factor X Mouse anti-Human, Alexa Fluor™ 350, Clone: MM0192-10A10, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP212198AF350
Catalog No. NP212198A30
Add to cart
Description
Description
Coagulation Factor X Monoclonal antibody specifically detects Coagulation Factor X in Human samples. It is validated for Western Blot.
Specifications
Specifications
Coagulation Factor X
MM0192-10A10
Western Blot
coagulation factor X, EC 3.4.21, EC 3.4.21.6, factor Xa, FX, FXA, prothrombinase, Stuart factor, Stuart-Prower factor
Mouse
IgG1
Store at 4°C in the dark.
Monoclonal
Human
Western Blot
Alexa Fluor™ 350
50 mM sodium borate with 0.05% sodium azide
F10
Human Factor X
0.1 mL
Primary
2159