Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Coagulation Factor X Rabbit anti-Human, Clone: 58, Novus Biologicals™
Rabbit Monoclonal Antibody
$321.00 - $493.00
Specifications
Antigen | Coagulation Factor X |
---|---|
Clone | 58 |
Dilution | Sandwich ELISA Detection 1:1000-1:10000 |
Applications | ELISA |
Classification | Monoclonal |
Description
Coagulation Factor X Monoclonal specifically detects Coagulation Factor X in Human samples. It is validated for Sandwich ELISA Detection.Specifications
Coagulation Factor X | |
Sandwich ELISA Detection 1:1000-1:10000 | |
Monoclonal | |
Purified | |
RUO | |
0.2 um filtered solution in PBS | |
2159 | |
IgG | |
Protein A purified |
58 | |
ELISA | |
Unconjugated | |
Rabbit | |
Human | |
coagulation factor X, EC 3.4.21, EC 3.4.21.6, factor Xa, FX, FXA, prothrombinase, Stuart factor, Stuart-Prower factor | |
Recombinant Human Coagulation Factor X Protein | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title