Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Coagulation Factor XI Rabbit anti-Human, Clone: 7, Novus Biologicals™
Rabbit Monoclonal Antibody
$321.00 - $493.00
Specifications
Antigen | Coagulation Factor XI |
---|---|
Clone | 7 |
Dilution | ELISA 1:5000-1:10000 |
Applications | ELISA |
Classification | Monoclonal |
Description
Coagulation Factor XI Monoclonal specifically detects Coagulation Factor XI in Human samples. It is validated for ELISA.Specifications
Coagulation Factor XI | |
ELISA 1:5000-1:10000 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
coagulation factor XI, EC 3.4.21, EC 3.4.21.27, FXIPlasma thromboplastin antecedent, MGC141891, PTA | |
Recombinant Human Coagulation Factor XI protein | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
7 | |
ELISA | |
Unconjugated | |
Rabbit | |
Prostate Cancer | |
0.2 um filtered solution in PBS | |
2160 | |
IgG | |
Protein A purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title